Order Entry
Belgium
ContactUsLinkComponent
HIV-2 gp32
HIV-2 gp32
Supplier:  Biorbyt
Certificates
HIV-2 gp32
Supplier:  Biorbyt

Specifications

  • Conjugation:
    Unconjugated
  • Storage conditions:
    Store at−20 °C.
  • Protein/peptide name:
    HIV-2 gp32
  • Purity:
    Greater than 95% by SDS-PAGE and RP-HPLC
  • Formulation:
    Liquid. Supplied in 20 mM PBS pH 7.8, 0.5 M NaCl, 1mM DTT, 8 M urea and 0.4 M imidazole.
  • Tested applications:
    WB , SDS-PAGE

Specifications

Related Information

Frequently Bought Together