Saisie de commande
Belgium
ContactUsLinkComponent
SARS-ACSMS
SARS-ACSMS
Catalog # BIRBORB97847-100
Supplier:  Biorbyt
CAS Number:  
SARS-ACSMS
Catalog # BIRBORB97847-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Cdt:
    100 µG
  • Conditions de stockage:
    Store at−20 °C.
  • Nom de la protéine/du peptide:
    SARS-ACSMS
  • Purity:
    Greater than 95% by SDS-PAGE
  • Molecular weight:
    37 kDa
  • Formulation:
    Liquid. Supplied in 25 mM Tris-HCl, 0.4% sarcosyl,0,25% Triton X-100 and 50% glycerol.
  • Tested applications:
    ELISA , WB

Specifications

About this item