Order Entry
Belgium
ContactUsLinkComponent
Mouse BD2 (from E. coli)
Mouse BD2 (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse BD2 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    Determined by its ability to chemoattract immature human dendritic cells using a concentration of 10.0-100.0 ng/ml. Less than 1EU/ug of rMuBD-2 as determined by LAL method.
  • Protein/peptide name:
    BD2
  • Purity:
    >98% pure by SDS-PAGE and HPLC analyses.
  • Molecular weight:
    5.54 kDa
  • Sequence:
    AVGSLKSIGY EAELDHCHT N GGYCVRAICP PSARRPGSCF PEKNPCCKYM K
  • Formulation:
    Sterile Filtered White lyophilized (freeze-dried) powder. We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom.
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together