Kallikrein (from plasma)
Catalog # BIRBORB82140-50
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:50 µG
- Source:Plasma
- Protein synonyms:Goat Serum Against Alpha-2 Antiplasmin
- Protein/peptide name:Kallikrein
- Purity:Purified.
- Formulation:Lyophilised.
- Tested applications:SDS-PAGE