Order Entry
Belgium
ContactUsLinkComponent
Human lipoprotein, high density (HDL) 2 subfraction (from plasma)
Human lipoprotein, high density (HDL) 2 subfraction (from plasma)
Catalog # BIRBORB82287-1
Supplier:  Biorbyt
CAS Number:  
Human lipoprotein, high density (HDL) 2 subfraction (from plasma)
Catalog # BIRBORB82287-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
    Plasma
  • Species:
    Human
  • Protein synonyms:
    Lipoprotein
  • Protein/peptide name:
    Lipoprotein, High Density (HDL) 2 Subfraction
  • Purity:
    Purified.
  • Formulation:
    Preservative: 1 mm Sodium azide

Specifications

About this item