Order Entry
Belgium
ContactUsLinkComponent
Human fibronectin (from plasma)
Human fibronectin (from plasma)
Catalog # BIRBORB82181-1
Supplier:  Biorbyt
CAS Number:  
Human fibronectin (from plasma)
Catalog # BIRBORB82181-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
    Plasma
  • Species:
    Human
  • Protein synonyms:
    CIG
  • Protein/peptide name:
    Fibronectin
  • Purity:
    Purified.
  • Formulation:
    Not lyophilised.
  • Tested applications:
    SDS-PAGE

Specifications

About this item