Order Entry
Belgium
ContactUsLinkComponent
Recombinant SARS-associated coronavirus (SARS-CoV) M protein (from E. coli)
Recombinant SARS-associated coronavirus (SARS-CoV) M protein (from E. coli)
Catalog # BIRBORB82444-1
Supplier:  Biorbyt
CAS Number:  
Recombinant SARS-associated coronavirus (SARS-CoV) M protein (from E. coli)
Catalog # BIRBORB82444-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Protein synonyms:
    SARS-Associated Coronavirus (SARS-CoV) M
  • Protein/peptide name:
    SARS-Associated Coronavirus (SARS-CoV) M protein
  • Purity:
    Purified.
  • Formulation:
    Not lyophilised.
  • Tested applications:
    ELISA , WB

Specifications

About this item