Recombinant Borrelia afzelii (from broth)
Catalog # BIRBORB82422-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:Broth
- Protein/peptide name:Borrelia afzelii
- Purity:Lysate.
- Formulation:Not lyophilised.
- Tested applications:ELISA , WB