Recombinant heat shock protein-70, Mycobacterium tuberculosis (from E. coli)
N° de catalogue BIRBORB82533-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Protein synonyms:Heat Shock-70
- Protein/peptide name:Heat Shock Protein-70, Mycobacterium tuberculosis
- Purity:Purified.
- Formulation:Lyophilised.
- Tested applications:ELISA , WB