Order Entry
Belgium
ContactUsLinkComponent
Recombinant herpes simplex virus-1 (HSV-1) glycoprotein D glycosylated (from P. pastoris)
Recombinant herpes simplex virus-1 (HSV-1) glycoprotein D glycosylated (from P. pastoris)
Catalog # BIRBORB82583-1
Supplier:  Biorbyt
CAS Number:  
Recombinant herpes simplex virus-1 (HSV-1) glycoprotein D glycosylated (from P. pastoris)
Catalog # BIRBORB82583-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    P. pastoris
  • Protein/peptide name:
    Herpes Simplex Virus-1 (HSV-1) Glycoprotein D Glycosylated
  • Purity:
    Purified.
  • Formulation:
    Not lyophilised.
  • Tested applications:
    ELISA , WB

Specifications

About this item