Treponema pallidum recombinant Treponemal membrane protein A (from Escherichia coli)
BIRBORB82574-100
: Biorbyt
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:T. pallidum
- Protein synonyms:TmpA|Treponema pallidum
- Protein/peptide name:Treponemal membrane Protein A
- Purity:Purified.
- Formulation:Not lyophilised.
- Tested applications:ELISA , WB , LF