Order Entry
Belgium
ContactUsLinkComponent
Human TNF alpha, Mutant (from E. coli)
Human TNF alpha, Mutant (from E. coli)
Supplier:  Biorbyt
Certificates
Human TNF alpha, Mutant (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The ED50 as determined by the cytolysis of murine L929 cells in the presence of Actinomycin is 0.05ng/ml, corresponding to Specific Activity of 100,000,000 IU/mg.
  • Protein synonyms:
    APC1
  • Protein/peptide name:
    TNF alpha, Mutant
  • Purity:
    > 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Tumor Necrosis Factor-alpha Variant in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MRKRKPVAHV VANPQAEGQL QWLNRRANAL LANGVELRDNQLVVPSEGLY LIYSQVLFKG QGCPSTHVLL THTISRIAVS YQTKVNLLSA IKSPCQRETP EGAEAKPWYE PIYLGGVFQL EKGDRLSAEI NRPDYLDFAE SGQVYFGIIAF.
  • Formulation:
    The protein was lyophilized after extensive dialysis against 0.5x PBS pH -7.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together