Rat SDF1 beta (from E. coli)
: Biorbyt
- Source:E. coli
- Species:Rat
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The specific activity as determined by its ability to chemoattract human peripheral blood monocytes at 100-150 ng/ml.
- Protein synonyms:C-X-C motif chemokine 12
- Protein/peptide name:SDF1 beta
- Purity:> 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized SDF-1b in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKSNNRQVC IDPKLKWIQE YLDKALNKRL KM.
- Formulation:The Rat SDF1B protein was lyophilized from concentrated (1 mg/ml) sterile solution containing 20 mM Phsophate buffer pH-7.4 and 0.15M NaCl.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together