Human recombinant IL22 (fromE. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Human
- Conditions de stockage:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Activité biologique:The Biological Activity was determined by the ability to activiate STAT following receptor ligand interaction.
- Synonyme de protéine:IL 22
- Nom de la protéine/du peptide:IL22
- Purity:> 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubilité:It is recommended to reconstitute the lyophilized Interleukin -22 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first five N-terminal amino acids was determined and was found to be Met-Ala-Pro-Ile-Ser.
- Formulation:Each mg contains 1.6mg mannitol and 1.2mg HSA.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together