Order Entry
Belgium
ContactUsLinkComponent
Human recombinant IL1RA (fromE. coli)
Human recombinant IL1RA (fromE. coli)
Supplier:  Biorbyt
Certificates
Human recombinant IL1RA (fromE. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The ED50 as determined by the dose-dependant inhibition of IL-1 stimulation of D10S cells was found to be 0.5 ng/ml.
  • Protein synonyms:
    DIRA
  • Protein/peptide name:
    IL1RA
  • Purity:
    > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleukin-1ra in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Met-Arg-Pro-Ser-Gly.
  • Formulation:
    Lyophilized protein with no additives.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together