Order Entry
Belgium
ContactUsLinkComponent
 
Mouse IL1 alpha (from E. coli)
 :  Biorbyt
 
Mouse IL1 alpha (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The ED50 as determined by the dose-dependant stimulation of murine D10S cells is 0.01 ng/ml, corresponding to Specific Activity of 100,000,000 IU/mg.
  • Protein synonyms:
    Hematopoietin 1
  • Protein/peptide name:
    IL1 alpha
  • Purity:
    > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleukin 1a in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Ser-Ala-Pro-Tyr-Thr.
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) sterile solution containing 1X PBS, pH=7.4.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

 

 

Frequently Bought Together