Order Entry
Belgium
ContactUsLinkComponent
Human IL1 alpha (from E. coli)
Human IL1 alpha (from E. coli)
Supplier:  Biorbyt
Certificates
Human IL1 alpha (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The ED50 as determined by the dose-dependant stimulation of murine D10S cells is 0.001 ng/ml, corresponding to Specific Activity of x 1,000,000,000 IU/mg.
  • Protein synonyms:
    Hematopoietin 1
  • Protein/peptide name:
    IL1 alpha
  • Purity:
    > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleukin alpha in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    SAPFSFLSNVKYNFMRIIKYEFILNDALNQSIIRANDQYLTAAALHNLDEAV KFDMGAYKSSKDDAKITVILRISKTQLYVTAQDEDQPVLLKEMPEIPKTITG SETNLLFFWETHGTKNYFTSVAHPNLFIATKQDYWVCLAGGPPSITDFQILE NQA.
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) sterile solution containing 20 mM Tris-HCL, pH=8, 5mM MgCl2 and 10% glycerol.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together