Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Cystatin-A (from E. coli)
Human recombinant Cystatin-A (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant Cystatin-A (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
  • Protein/peptide name:
    Cystatin-A
  • Formulation:
    Cystatin-A in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together