Human recombinant LBP (from CHO cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Type de protéine/peptide:Recombinant
- Source:CHO cells
- Species:Human
- Conditions de stockage:Lyophilized LBP Human Recombinant although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution LBP should be stored at 4 °C between 2-7 days and for future use below −18 °C. Please prevent freeze/thaw cycles.
- Activité biologique:Up to 0.2 ug/ ml LBP mediates binding of FITC-LPS (0.5ug/ml) to CD14C HO transfectants (FACS).
- Synonyme de protéine:Lipopolysaccharide-binding
- Nom de la protéine/du peptide:LBP
- Solubilité:Every 10ug of recombinant human LBP should be reconstituted using 40ul of sterile H2O.
- Molecular weight:On SDS-PAGE commassie blue stained gel, the 95% purified recombinant protein shows band at 58 kDa.
- Formulation:Recombinant Human LBP was lyophilized from protein solution (0.25 mg/ml) containing phosphate-buffered saline, pH 7.2.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together