Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant mCD14 (from CHO cells)
Mouse recombinant mCD14 (from CHO cells)
Supplier:  Biorbyt
Certificates
Mouse recombinant mCD14 (from CHO cells)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized Mouse CD14 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Mouse CD14 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Protein synonyms:
    Monocyte differentiation antigen CD14
  • Protein/peptide name:
    mCD14
  • Solubility:
    It is recommended to reconstitute the lyophilized Mouse CD14 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Formulation:
    CD14 was lyophilized from concentrated protein solution (1 mg/ml) containing phosphate-buffered saline, pH 7.2.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together