Order Entry
Belgium
ContactUsLinkComponent
Human recombinant ZA2G (from HEK293 cells)
Human recombinant ZA2G (from HEK293 cells)
Supplier:  Biorbyt
Certificates
Human recombinant ZA2G (from HEK293 cells)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilized ZA2G although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution ZA2G should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please avoid freeze/thaw cycles.
  • Biological activity:
    Differentiated human SGBS adipocytes were incubated for 18 at two dose levels of rhZA2G 5 and 20 ug/ml. Lipolysis was quantified by measuring glycerol release into the medium using standard protocol. Isoproterenol (10 uM) and IBMX (100 uM) were used as positive controls. "Con" stands for the negative control. There was 3-fold increase in glycerol release with both doses. The increase was statistically significant at ug/ml dose of rhZA2G (p<0.01) as well as in positive controls.
  • Protein synonyms:
    Zn-alpha-2-glycoprotein
  • Protein/peptide name:
    ZA2G
  • Purity:
    Greater than 90.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
  • Solubility:
    Add deionized water to working concentration approximately 0.5 mg/ml and let the lyophilized pellet dissolve completely.
  • Sequence:
    ASWSHPQFEK GSQENQDGRY SLTYIYTGLS KHVEDVPAFQ ALGSLNDLQF FRYNSKDRKS QPMGLWRQVEGMEDWKQDSQ LQKAREDIFM ETLKDIVEYY NDSNGSHVLQ GRFGCEIENN RSSGAFWKYY YDGKDYIEFNKEIPAWVPFD PAAQITKQKW EAEPVYVQRA KAYLEEECPA TLRKYLKYSK NILDRQDPPS VVVTSHQAPG EKKKLKCLAYDFYPGKIDVH WTRAGEVQEP ELRGDVLHNG NGTYQSWVVV AVPPQDTAPY SCHVQHSSLA QPLVVPWEAS.
  • Formulation:
    Filtered (0.4 um) and lyophilized in 0.5 mg/ml in 0.1M Tris-HCl pH 8.0 and 150mM NaCl.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together