Order Entry
Belgium
ContactUsLinkComponent
LLO (from E. coli)
LLO (from E. coli)
Supplier:  Biorbyt
Certificates
LLO (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Storage conditions:
    LLO although stable at room temperature for week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.
  • Biological activity:
    LLO has hemolytic activity 1.25x106 HU/mg.2mM DTT is used to reactivate the toxin.
  • Protein synonyms:
    Listeriolysin-O
  • Protein/peptide name:
    LLO
  • Purity:
    Greater than 90.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Formulation:
    The protein contains 50mM NaH2PO4, 1mM EDTA, 2.7mM KCl, 1mM DTT, 5% glycerol and 0.5M NaCl.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together