Order Entry
Belgium
ContactUsLinkComponent
Human recombinant PI3Kb (from insect cells (Sf9))
Human recombinant PI3Kb (from insect cells (Sf9))
Supplier:  Biorbyt
Certificates
Human recombinant PI3Kb (from insect cells (Sf9))
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Storage conditions:
    PI3Kb although stable at 4 °C for week, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
  • Biological activity:
    ~ nmol/mg/min using phosphatidylinositol as the substrate.
  • Protein synonyms:
    Phosphoinositide 3-kinase beta p110d
  • Protein/peptide name:
    PI3Kb
  • Purity:
    Greater than 90.0% as determined by SDS Page.
  • Formulation:
    0.5 mg/ml solution in 10mM Hepes, pH 7.5, 100mM NaCl, 2.5mM MgCl2, and 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together