Human VEGF receptor 1 D1-5 (from Insect cells)
: Biorbyt
- Source:Insect cells
- Species:Human
- Storage conditions:Lyophilized FLT-1 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution FLT1 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Biological activity:The activity of FLT1 D5 was determined by its ability to abolish the binding of iodinated VEGF to solid surfaces or cell surfaces. The ED50 for this effect is typically 10 ng/ml. In 13 day CAM-assay sVEGFR-1 is able to inhibit VEGF stimulated sprouting of capillaries at 30 pM.
- Protein synonyms:Vascular endothelial growth factor A
- Protein/peptide name:VEGF receptor 1 D1-5
- Purity:Greater than 90.0% as determined by(a)Analysis by RP-HPLC.(b)Analysis by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized FLT1 D5 in sterile water not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:FLT1 D1-5 was lyophilized from concentrated (1 mg/ml) sterile solution containing no additives.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together