HIV-1 nef Clade B (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Storage conditions:Protein is shipped at ambient temperature. Upon arrival, Store at −20 °C.
- Protein/peptide name:HIV-1 nef Clade B
- Purity:Greater than 90.0% as determined by HPLC analysis and SDS-PAGE.
- Solubility:Reconstitute with PBS in apirogenic water or buffer with minimum ionic force to guarantee the correct folding of Nef protein.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together