Order Entry
Belgium
ContactUsLinkComponent
Human recombinant GDF3
Human recombinant GDF3
Supplier:  Biorbyt
Certificates
Human recombinant GDF3
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    Determined by its ability to inhibit induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The ED50 for this effect is 100-150 ng/ml.
  • Protein synonyms:
    Growth
  • UniProtKB:
    Q9NR23
  • Protein/peptide name:
    GDF3
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized GDF3 in sterile 100mM Acetate buffer pH-4 at concentration of 0.5mg/ml. For the dilution into higher pH values, it is recommended to dilute the protein to concentration of 10ug/ml. Please note that in high
  • Sequence:
    AAIPVPKLSCKNLCHRHQLFINFRDLGWHKWIIAPKGFMANYCHGECPFSLTISLNSSNYAFMQALMHAVDPEIPQAVCIPTKLSPISMLYQDNNDNVILRHYEDMVVDECGCG
  • Formulation:
    The protein was lyophilized from concentrated solution (0.5mg/ml) containing 30mM Acetate buffer pH-4.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together