Rat recombinant IL15
: Biorbyt
- Protein/peptide type:Recombinant
- Species:Rat
- Storage conditions:Store at −20 °C.
- Biological activity:The ED50 as determined by the dose-dependant stimulation of the proliferation of Mouse CTLL-2 cells is < 10 ng/ml, corresponding to a specific activity of > 1 x 105 units/mg.
- Protein synonyms:IL-T
- UniProtKB:P97604
- Protein/peptide name:IL15
- Purity:>98%
- Solubility:It is recommended to reconstitute the lyophilized Interleukin-15 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:MNWIDVRYDL EKIESLIQFI HIDTTLYTDS DFHPSCKVTA MNCFLLELQV ILHEYSNMTL NETVRNVLYL ANSTLSSNKN VIESGCKECE ELEERNFTEF LQSFIHIVQM FINTS
- Formulation:The protein was lyophilized from 10mM Tris, pH-8.5.
- Tested applications:SDS-PAGE
Frequently Bought Together