Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant prolactin
Mouse recombinant prolactin
Supplier:  Biorbyt
Certificates
Mouse recombinant prolactin
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    Determined by its ability to induce the proliferation of rat Nb2-11 cells in the concentration range of 0.1-1.0 ng/ml.
  • Protein synonyms:
    Mammotropin
  • UniProtKB:
    P06879
  • Protein/peptide name:
    Prolactin
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized Prolactin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MLPICSAGDC QTSLRELFDR VVILSHYIHT LYTDMFIEFD KQYVQDREFM VKVINDCPTS SLATPEDKEQ ALKVPPEVLL NLILSLVQSS SDPLFQLITG VGGIQEAPEY ILSRAKEIEE QNKQLLEGVE KIISQAYPEA KGNGIYFVWS QLPSLQGVDE ESKILSLRNT IRCLRRDSHK VDNFLKVLRC QIAHQNNC
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) solution with 10mM sodium Phosphate buffer pH=8 and 50mM Nacl.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together