Mouse stem cell factor
Supplier: Biorbyt
Certificates
About this item
Specifications
- Species:Mouse
- Storage conditions:Store at −20 °C.
- Biological activity:The ED50 as determined by the dose-dependent stimulation of the proliferation of the human TF-1 cells is < 10 ng/ml, corresponding to a specific activity of > 1 x 105 units/mg.
- Protein synonyms:Kit ligand Precursor
- UniProtKB:P20826
- Protein/peptide name:Stem Cell Factor
- Purity:>98%
- Solubility:It is recommended to reconstitute the lyophilized Stem Cell Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
- Formulation:Lyophilized from concentrated (1mg/ml) solution in water containing 10mM Sodium phosphate buffer pH=6.5.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together