Order Entry
Belgium
ContactUsLinkComponent
Human recombinant I 309
Human recombinant I 309
Supplier:  Biorbyt
Certificates
Human recombinant I 309
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    Determined by its ability to chemoattract total human T cell population using a concentration range of 10.0-100.0 ng/ml.
  • Protein synonyms:
    CCL1
  • UniProtKB:
    P22362
  • Protein/peptide name:
    I 309
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized I-309 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    SKSMQVPFSR CCFSFAEQEI PLRAILCYRN TSSICSNEGL IFKLKRGKEA CALDTVGWVQ RHRKMLRHCP SKRK
  • Formulation:
    The CCL1 protein was lyophilized with no additives.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together