Order Entry
Belgium
Orders LinkContactUsLinkComponent
ORP electrode

ORP electrode

ORP electrode, mV ± 2000, GE 105-BNC, BNC, 1 m cable, 3 mol/l KCl, 0...+80 °C, 12×120 mm
Catalog # GRSG607798
Supplier:   Greisinger
ORP electrode
Catalog # GRSG607798
Supplier:  Greisinger
ORP electrode
ORP electrode

ORP electrode, mV ± 2000, GE 105-BNC, BNC, 1 m cable, 3 mol/l KCl, 0...+80 °C, 12×120 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Redox electrode with BNC plug.

Specifications
  • Connection:
    BNC, 1 m cable
  • Ø×L:
    12×120 mm
  • Type:
    GE 105-BNC
  • Description:
    ORP electrode, mV ± 2000
  • Electrolyte:
    3 mol/l KCl
  • Temp.:
    0...+80 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.