Orders LinkContactUsLinkComponent
Lockout stations, adjustable
Lockout stations, adjustable
Supplier:   Brady
Lockout stations, adjustable
Supplier:  Brady
Certificates
Lockout stations, adjustable
Lockout stations, adjustable
3 Options Available Below
Product Details & Documents
These lockout stations (without components) are constructed of sturdy white PS with pre-drilled holes for easy wall mounting.
  • Pegboard-style holes for hooks drilled into back (4 hooks supplied standard)
  • Built-in lock rail holds a different number of locks depending on lock size
Small station: Holds 10 to 15 locks.
Large station: Holds 15 to 20 locks.
Extra-large station: Holds 20 to 25 locks.
Ordering information: Lockout components need to be ordered seperately.
Product Family Options
Product Information
  • Type
    Colour
    W×D×H
    Availability
    Price
  • Lockout station, small
    White
    311×97×591 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 197,00 €
    197,00 €
    1 * 1 ST
       197,00 €
  • Lockout station, large
    White
    495×127×660 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 374,00 €
    374,00 €
    1 * 1 ST
       374,00 €
  • Lockout station, extra large
    White
    635×133×762 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 521,00 €
    521,00 €
    1 * 1 ST
       521,00 €