ContactUsLinkComponent
Avantor® Alltima, HPLC Columns, Amino
Avantor® Alltima, HPLC Columns, Amino
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Avantor® Alltima, HPLC Columns, Amino
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • HICH88217
  • 88217RP
  • HICH88218
  • AL-5NH-250DF
  • HICH81183
  • HICH81190
  • 96085/N
  • HICH88205
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Surface area:
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • USP code:
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • L8
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)