Order Entry
Belgium
ContactUsLinkComponent
VWR®, UV/Visible spectroscopy linearity standards at 235, 257, 313 and 350 nm
VWR®, UV/Visible spectroscopy linearity standards at 235, 257, 313 and 350 nm
Catalog # 84756.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, UV/Visible spectroscopy linearity standards at 235, 257, 313 and 350 nm
Catalog # 84756.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Blank - 0,001 M perchloric acid for linearity standards at 235, 257, 313 and 350 nm
  • Seq:
    34

Specifications

About this item

Linearity standards packed in permanently sealed UV cuvettes or 100 ml amber bottles.