ContactUsLinkComponent
Replacement needle and plunger kit (SGE) for PerkinElmer autosampler syringes
Catalog # REST24818
Supplier:  Restek
undefined
Replacement needle and plunger kit (SGE) for PerkinElmer autosampler syringes
Catalog # REST24818
Supplier:  Restek
Specifications
  • Gauge:
    23
  • Type of tip:
    Conical needle
  • Description:
    Replacement needle and plunger kit, 0,5 µl
  • Length:
    70 mm
  • Pk:
    1