Order Entry
Belgium
ContactUsLinkComponent
Sodium chloride dishwasher salt, ProCare Universal 61 for water softening
Sodium chloride dishwasher salt, ProCare Universal 61 for water softening
Catalog # MIEL9195780
Supplier:  MIELE
CAS Number:  
undefined
Sodium chloride dishwasher salt, ProCare Universal 61 for water softening
Catalog # MIEL9195780
Supplier:  MIELE
CAS Number:  

Specifications

  • Pk:
    6 kg
  • Seq:
    4122

Specifications

About this item

 :
 :58.44 g/mol
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477