Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chromatography Columns
Glass column for column elution
Glass column for column elution
Glass column for column elution
Catalog # BEHRB00231897
Supplier:
BEHR
Expand image
Glass column for column elution
Catalog # BEHRB00231897
Supplier:
BEHR
Specifications
Ø int.:
60,00 mm
Capacity:
950 ml
Description:
Glass column with GL45 on both ends, GS 950, according to LUA bulletin no. 20
Length:
320 mm
Pk:
1
Specifications
About this item
Product Details
Product Details