ContactUsLinkComponent
Red detection kit (GFP-CERTIFIED®), LYSO-ID®
Red detection kit (GFP-CERTIFIED®), LYSO-ID®
Catalog # ENZOENZ510050100
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Red detection kit (GFP-CERTIFIED®), LYSO-ID®
Catalog # ENZOENZ510050100
Supplier:  Enzo Life Sciences
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Red detection kit (GFP-CERTIFIED®), LYSO-ID®
  • Seq:
    02
Specifications
About this item
LYSO-ID® Red Detection Kit (GFP-CERTIFIED®) is an acidic organelle-selective dye for live cell staining of lysosomes. It is an ideal choice for fluorescence co-localisation imaging with GFP-tagged proteins and providing spectrally pure red signal.
  • No photo conversion to a green emitting dye
  • No complicating metachromatic dual emission artifacts, facilitating analysis with GFP, FITC and other fluorescent probes
  • Highly resistant to photobleaching and concentration quenching
  • Ensures strong and consistent fluorescence signal, even after extended viewing periods
LYSO-ID® Red Detection Kit is for use with GFP-expressing cell lines, as well as cells expressing blue, cyan or yellow fluorescent proteins.
Delivery: Kit contains LYSO-ID® Red Detection Reagent, Hoechst 33342 nuclear stain, chloroquine control and 10X assay buffer.