ContactUsLinkComponent
Blue light LED transilluminators, Syngene UltraBright-LED
Blue light LED transilluminators, Syngene UltraBright-LED
Catalog # SYNOGX-ULTRABRIGHT
undefined
Blue light LED transilluminators, Syngene UltraBright-LED
Catalog # SYNOGX-ULTRABRIGHT
Specifications
  • Wavelength:
    470 nm
  • Description:
    Blue light LED transilluminator, UltraBright-LED
  • Dimensions:
    330×280×90 mm
  • Pk:
    1
Specifications
About this item
Blue light transilluminators are often used as an alternative to UV transilluminators when users wish to use ‘safe dyes’ instead of ethidium bromide. A blue light transilluminator is a ‘safe’ light source, in that the user is not exposed to harmful UV radiation and samples are free from photonicking. UltraBright-LED transilluminator uses side mounted 470 nm LED arrays for illumination and can be used with gels up to 20×16 cm.