Orders LinkContactUsLinkComponent
Samplers, MultiThief
Samplers, MultiThief
Samplers, MultiThief
Samplers, MultiThief
Samplers, MultiThief
The MultiThief is designed for the multi-level sampling of free flowing powders and granules.
  • Variable sample volumes up to 60 ml
  • Suitable for free fl owing powders and granules
  • Can be quickly and easily changed by removing cells
Product Family Options
  • Description
    Material
    Length
    No. of chambers
    Ø
    Volume
  • Sampler
    Stainless steel 316
    1000 mm
    3
    40 mm
    1 * 1 ST
    1 * 1 ST - 1 320,00 €
    1 320,00 €
    1 * 1 ST
       1 320,00 €
  • Sampler
    Stainless steel 316L
    1500 mm
    4
    40 mm
    1 * 1 ST
    1 * 1 ST - 1 560,00 €
    1 560,00 €
    1 * 1 ST
       1 560,00 €
  • Sampler
    Stainless steel 316
    2000 mm
    4
    40 mm
    1 * 1 ST
    1 * 1 ST - 1 630,00 €
    1 630,00 €
    1 * 1 ST
       1 630,00 €
Inserts
  • Description
    Material
    Length
    No. of chambers
    Ø
    Volume
  • Blank insert for MultiThief
    Stainless steel 316L
    1 * 1 ST
    1 * 1 ST - 308,00 €
    308,00 €
    1 * 1 ST
       308,00 €
  • Insert for MultiThief
    Stainless steel 316L
    5 ml
    1 * 1 ST
    1 * 1 ST - 327,00 €
    327,00 €
    1 * 1 ST
       327,00 €
  • Insert for MultiThief
    Stainless steel 316L
    10 ml
    1 * 1 ST
    1 * 1 ST - 327,00 €
    327,00 €
    1 * 1 ST
       327,00 €
  • Insert for MultiThief
    Stainless steel 316L
    25 ml
    1 * 1 ST
    1 * 1 ST - 327,00 €
    327,00 €
    1 * 1 ST
       327,00 €
  • Insert for MultiThief
    Stainless steel 316L
    50 ml
    1 * 1 ST
    1 * 1 ST - 327,00 €
    327,00 €
    1 * 1 ST
       327,00 €
  • Insert for MultiThief
    Stainless steel 316L
    60 ml
    1 * 1 ST
    1 * 1 ST - 327,00 €
    327,00 €
    1 * 1 ST
       327,00 €