Mouse recombinant WNT3A
ENZOENZ60001C001
:
- Pk:1 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Seq:02
- Reconstitution instructions:Prepare 5 μg/ml stock solution of Wnt3a by adding 200 μl deionised water into the vial of lyophilised Wnt3a
- Protein/peptide name:WNT3A
- Purity:>80%
- Molecular weight:~40 kDa
- Formulation:Lyophilised solid
Wnt expression and signal transduction have been identified as critical components in regulating embryogenesis, cell proliferation, and bone, glucose and lipid metabolism. Wnt-3a signalling also plays a role in regulating the differentation of stem cells. Wnt-3a is a member of the highly conserved, structurally related cysteine-rich Wnt family glycoproteins important for normal developmental processes and oncogenesis.
- Produced in CHO cells
- Purified by SDS-PAGE
- Endotoxin content determined by LAL method
Wnt-3a can induce β-catenin mediated response in a TCF/LEF reporter gene luciferase assay.