Order Entry
Belgium
ContactUsLinkComponent
Alginate spatula
Alginate spatula
Catalog # HAMMHWN271-21
Supplier:  HAMMACHER KARL
undefined
Alginate spatula
Catalog # HAMMHWN271-21
Supplier:  HAMMACHER KARL

Specifications

  • Length:
    210 mm
  • Seq:
    07
  • Environmentally Preferable:
  • Blade width:
    30 mm
  • Pk:
    1

Specifications

About this item

Alginate spatula with wooden handle and blade made of WIRONIT steel.