ContactUsLinkComponent
Beakers, heavy duty, Berzelius, tall form, borosilicate glass, PYREX®
Beakers, heavy duty, Berzelius, tall form, borosilicate glass, PYREX®
Catalog # BIBB1004-250
Supplier:  SciLabware
Supplier #: 1004-250
undefined
Beakers, heavy duty, Berzelius, tall form, borosilicate glass, PYREX®
Catalog # BIBB1004-250
Supplier:  SciLabware
Supplier #: 1004-250
Log in for availability
Select an Option
60 mm
Select an Option
250 ml
Select an Option
120 mm
Select an Option
02
Select an Option
10
About this item
Made of PYREX® borosilicate glass.
  • Thicker walls and heavier bases
  • Reinforced rims and spout, less prone to breakage during washing
  • Printed with Trace Code for downloadable batch certificate
These heavy duty beakers with white graduations and marking spot are designed for longer life and increased user safety.
Frequently Bought Together