Corning® Microplate Lids
About this item
All lids are made of rigid polystyrene except where indicated.
All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning® microplates.
The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
5 Options Available Below
Product Details & Documents
All lids are made of rigid polystyrene except where indicated.
All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning® microplates.
The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
Specifications for Product Family
Specifications
- Catalog No:
- 734-1538
- 734-1539
- 734-4165
- 734-1669
- 734-1668
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
Specifications
Product Family Options
Product Information
- SterilityAvailabilityPrice
- 734-1538Universal lid without corner notchSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-1538Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRECs, ISO 14001 - KennebunkLow Manufacturing Impact - 734-1539Universal lid with corner notchSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-1539Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRECs, ISO 14001 - KennebunkLow Manufacturing Impact - 734-4165Black universal lid with corner notchSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-4165Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRECs, ISO 14001 - KennebunkLow Manufacturing Impact - 734-1669Low evaporation lid with corner notch and condensation rings for 96-well microplates only (not 2 ml block)Low evaporation lid with corner notch and condensation rings for 96-well microplates only (not 2 ml block)SterileLog in for availability1 * 50 ST130,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-1669Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRECs, ISO 14001 - KennebunkLow Manufacturing Impact - 734-1668Low evaporation lid with corner notch and condensation rings for 96-well microplates only (not 2 ml block)Low evaporation lid with corner notch and condensation rings for 96-well microplates only (not 2 ml block)SterileLog in for availability1 * 100 ST300,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-1668Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRECs, ISO 14001 - KennebunkLow Manufacturing Impact
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.