Order Entry
Belgium
ContactUsLinkComponent
Potassium dichromate 0.020 mol/l in aqueous solution, Titripur® for COD determination, Supelco®
Health
Potassium dichromate 0.020 mol/l in aqueous solution, Titripur® for COD determination, Supelco®
Catalog # 1.09119.1000
Supplier:  Merck
CAS Number:  
Health
Potassium dichromate 0.020 mol/l in aqueous solution, Titripur® for COD determination, Supelco®
Catalog # 1.09119.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :294.18 g/mol
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
UN:3287
 :GHS08
Storage Temperature:Ambient
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367