Order Entry
Belgium
ContactUsLinkComponent
Fluorescent calcium indicator, GFP-CERTIFIED® FLUOFORTE®
Fluorescent calcium indicator, GFP-CERTIFIED® FLUOFORTE®
Catalog # ENZOENZ525550250
Supplier:  Enzo Life Sciences
CAS Number:  
Fluorescent calcium indicator, GFP-CERTIFIED® FLUOFORTE®
Catalog # ENZOENZ525550250
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Description:
    Fluorescent calcium indicator reagent, GFP-CERTIFIED® FLUOFORTE®
  • Seq:
    02
  • Purity:
    ≥95 % (HPLC)
  • Molecular weight:
    800

Specifications

About this item

GFP-CERTIFIED® FLUOFORTE® is a fluorescent calcium indicator relative to other red fluorescent dyes.

Can be used for the detection of intracellular calcium mobilisation across a broad spectrum of biological targets.

Abs/Em.: 524/ 551 nm

Caution: Esterase hydrolysis is not required for live cell analysis via flow cytometry or fluorescence microscopy.