ContactUsLinkComponent
DNA methylation kit, express (shallow well)
DNA methylation kit, express (shallow well)
Catalog # ENZOENZ450030002
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
DNA methylation kit, express (shallow well)
Catalog # ENZOENZ450030002
Supplier:  Enzo Life Sciences
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Express DNA methylation kit (shallow well), 2×96 wells
  • Seq:
    02
Specifications
About this item
The Express DNA Methylation Kit (shallow well) features high-throughput (96-well) bisulfite treatment and conversion of DNA for methylation analysis.
  • Ready to use conversion reagent is added directly to DNA
  • Easy to follow instructions
  • High yield
DNA denaturation and bisulfite conversion processes are combined with added heat to facilitate rapid denaturation. Desulphonation and clean-up of the converted DNA is performed using a unique 96-well spin-plate. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
Delivery: Kit contains Express conversion reagent, Methylation binding buffer, Methylation wash buffer concentrate, L-Desulphonation reagent, Methylation elution buffer, Express binding plate (shallow), Express conversion plates, collection plates and elution plates.