GC Columns, Rt-bDEXm™
Supplier: Restek
Certificates
About this item
Separations of more than one hundred chiral compounds have been achieved using our unique DEX columns, and DEX columns continue to demonstrate stability after hundreds of temperature program cycles. Permethylated beta cyclodextrin added into 14% cyanopropylphenyl / 86% dimethyl polysiloxane.
Frequently Bought Together