Photoionisation lamps
: Restek
Photoionisation (PID) lamps are vacuum tubes designed to emit ultraviolet energy.
Model 108-10.0/10.6: Features both 10,0 and 10,6 eV potential, has a 0,781" base diameter and is used in Tracor, OI and Baseline instruments.
Model 103: Has a 34,9 mm base and is used in HNU and SRI detectors.
Model 108-BTEX: Lamp’s higher output makes it ideal for detection of BTEX compounds.
Frequently Bought Together