ContactUsLinkComponent
MSD conversion fitting
MSD conversion fitting
Catalog # REST21314
Supplier:  Restek
undefined
MSD conversion fitting
Catalog # REST21314
Supplier:  Restek
Specifications
  • Description:
    MSD conversion fitting
  • Material:
    Nickel plated brass
  • Pk:
    1
Specifications
About this item
This conversion fitting is a flat, soft aluminum sealing ring deforms and butt-seals against the MSD interface. A standard Vespel® ferrule seals the column and 1,59 mm stainless steel nut.
  • Fitting is constructed of nickel-plated brass for longevity and softness
  • Use any standard Vespel® or Vespel®/graphite 1,59 mm ferrule
  • Improved design reduces chance of leaks
Delivery: This fitting includes a 1,59 mm stainless steel nut and two replacement sealing rings. Ferrules have to be ordered separately.